Skip to Content
Merck
All Photos(1)

Key Documents

S60535

N(2),N(2),N(2),N(2),N(4),N(4),N(4),N(4),N(6),N(6),N(6),N(6)-DODECAETHYL-1,3,5,2LAMBDA(5),4LAMBDA(5),6LAMBDA(5)-TRIAZATRIPHOSPHININE-2,2,4,4,6,6-HEXAMINE

AldrichCPR

Sign Into View Organizational & Contract Pricing

Select a Size

S60535-100MG
€70.10

€70.10


Check Cart for Availability


Select a Size

Change View
S60535-100MG
€70.10

About This Item

Linear Formula:
C24H60N9P3
CAS Number:
Molecular Weight:
567.728
MDL number:

€70.10


Check Cart for Availability

Other Notes

Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.

NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service