Skip to Content
Merck
All Photos(1)

Key Documents

N21308

Sigma-Aldrich

3-Nitro-1,2-phenylenediamine

98%

Synonym(s):

1,2-Diamino-3-nitrobenzene, 3-Nitro-o-phenylenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H3(NH2)2
CAS Number:
Molecular Weight:
153.14
Beilstein:
909344
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

mp

156-159 °C (lit.)

SMILES string

Nc1cccc(c1N)[N+]([O-])=O

InChI

1S/C6H7N3O2/c7-4-2-1-3-5(6(4)8)9(10)11/h1-3H,7-8H2

InChI key

IOCXBXZBNOYTLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tamara Regina Calvo et al.
Evidence-based complementary and alternative medicine : eCAM, 2011, 323276-323276 (2009-08-22)
Indigofera truxillensis and I. suffruticosa, are used as a source of indigo dye and to treat several diseases. The mutagenic activity of the methanolic extracts from aerial parts, glycerolipid, flavonoid and alkaloid fractions of the extract were evaluated by means

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service