Key Documents
MT51340500
Mettler Toledo DX series ISE half-cells electrode
Fluoride selective
Select a Size
1 EA
€1,140.00
€1,140.00
Check Cart for Availability
Select a Size
Change View
1 EA
€1,140.00
About This Item
UNSPSC Code:
39121436
NACRES:
NB.79
€1,140.00
Check Cart for Availability
material
membrane (Solid state)
shaft (POM)
manufacturer/tradename
Mettler-Toledo 51340500
measuring range
1.0-1.0-4
temperature measuring range
0-80 °C
pH
4-10
General description
Modular and versatile the METTLER TOLEDO DX series ion-selective half-cells consist of two elements: a universal shaft and an ion-specific membrane module. This module may be exchanged allowing you to measure the ion of your choice. Just order the membrane kit specific for that ion, mount the new module onto the shaft of your ISE half-cell, and you have a new ISE!
electrolyte for reference electrode: 1 mol/L KNO3, ISA solution: 10 mol/L NaOH
Choose from one of the most recent versions:
Certificates of Analysis (COA)