Key Documents
MT30019823
Mettler-Toledo SevenCompact uPlace electrode holder, (included in standard delivery)
Synonym(s):
electrode stand
Select a Size
1 EA
€250.00
€250.00
Check Cart for Availability
Select a Size
Change View
1 EA
€250.00
About This Item
UNSPSC Code:
41115603
NACRES:
NB.79
€250.00
Check Cart for Availability
manufacturer/tradename
Mettler-Toledo 30019823
General description
  • Mettler-Toledo SevenCompact uPlace electrode holder is an accessory for use with the pH meter.
  • The electrode arm of the holder can be moved straight up and down and can be operated with one hand.
  • Works more efficiently when compared to the traditional designs.
Legal Information
SevenCompact is a trademark of Mettler-Toledo, Inc.
uPlace is a trademark of Mettler-Toledo, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)