Skip to Content
Merck
All Photos(1)

Documents

M5387

Sigma-Aldrich

p-Methoxybenzyl S-(4,6-dimethylpyrimidin-2-yl) thiocarbonate

powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H16N2O3S
CAS Number:
Molecular Weight:
304.36
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

powder

color

light yellow

storage temp.

2-8°C

SMILES string

COc1ccc(COC(=O)Sc2nc(C)cc(C)n2)cc1

InChI

1S/C15H16N2O3S/c1-10-8-11(2)17-14(16-10)21-15(18)20-9-12-4-6-13(19-3)7-5-12/h4-8H,9H2,1-3H3

InChI key

DDIJXDDZBQMPDQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service