Skip to Content
Merck
All Photos(1)

Documents

M15206

Sigma-Aldrich

5-Methoxy-2-methylbenzselenazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NOSe
CAS Number:
Molecular Weight:
226.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

refractive index

n20/D 1.639 (lit.)

bp

115-117 °C/1 mmHg (lit.)

density

1.491 g/mL at 25 °C (lit.)

SMILES string

COc1ccc2[se]c(C)nc2c1

InChI

1S/C9H9NOSe/c1-6-10-8-5-7(11-2)3-4-9(8)12-6/h3-5H,1-2H3

InChI key

LVIYTMFDCXRSDL-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service