Skip to Content
Merck
All Photos(1)

Documents

K2208

Sigma-Aldrich

Sodium mesoxalate monohydrate

Synonym(s):

Dihydroxymalonic acid disodium salt, Ketomalonic acid disodium salt, Mesoxalic acid disodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(HO)2C(COONa)2
CAS Number:
Molecular Weight:
180.02
Beilstein:
4924524
EC Number:
MDL number:
UNSPSC Code:
12352100

SMILES string

[Na+].[Na+].OC(O)(C([O-])=O)C([O-])=O

InChI

1S/C3H4O6.2Na/c4-1(5)3(8,9)2(6)7;;/h8-9H,(H,4,5)(H,6,7);;/q;2*+1/p-2

InChI key

HMBGDJCPFGQLIO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service