JKA10034
8arm-PEG20K
average MN 20,000, Vinylsulfone, tripentaerythritol core, thiol reactive
Synonym(s):
8arm-PEG-Vinylsulfone, tripentaerythritol core, 8arm-PEG-VS, Polyethylene glycol
About This Item
Recommended Products
product name
8arm-PEG20K-Vinylsulfone, tripentaerythritol core, average Mn 20,000
form
solid
mol wt
average Mn 20,000
reaction suitability
reactivity: thiol reactive
polymer architecture
shape: 8-arm
functionality: homofunctional
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Legal Information
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Immune system defends against disease by hydrolyzing foreign molecules; proteases, nucleases, liver, and kidneys are involved.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service