Skip to Content
Merck
All Photos(1)

Key Documents

GF81185272

Stainless Steel - AISI 321 alloy, FeCr18Ni9Ti

foil, 15mm disks, 0.2mm thickness, annealed

Synonym(s):

FE210310

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
11171600
NACRES:
NA.23
Pricing and availability is not currently available.

form

annealed
foil (disks)

manufacturer/tradename

Goodfellow 811-852-72

size × thickness

15 mm × 0.2 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF03977781GF11785057GF17565824
manufacturer/tradename

Goodfellow 811-852-72

manufacturer/tradename

Goodfellow 039-777-81

manufacturer/tradename

Goodfellow 117-850-57

manufacturer/tradename

Goodfellow 175-658-24

size × thickness

15 mm × 0.2 mm

size × thickness

8 mm × 0.25 mm

size × thickness

4 mm × 0.15 mm

size × thickness

50 mm × 0.125 mm

General description

For updated SDS information please visit www.goodfellow.com.
Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service