Skip to Content
Merck
All Photos(1)

Documents

F5312

Sigma-Aldrich

Fmoc-(R)-propargyl-Ala-OH

Synonym(s):

(R)-N-Fmoc-α-propargyl-Ala-OH, (R)-N-Fmoc-α-propargylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H19NO4
CAS Number:
Molecular Weight:
349.38
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: click chemistry

application(s)

peptide synthesis

functional group

Fmoc

shipped in

wet ice

storage temp.

−20°C

SMILES string

O=C(N[C@](C)(CC#C)C(O)=O)OCC1C2=CC=CC=C2C3=C1C=CC=C3

InChI

1S/C21H19NO4/c1-3-12-21(2,19(23)24)22-20(25)26-13-18-16-10-6-4-8-14(16)15-9-5-7-11-17(15)18/h1,4-11,18H,12-13H2,2H3,(H,22,25)(H,23,24)/t21-/m1/s1

InChI key

ZXOKSWZUJXKQCQ-OAQYLSRUSA-N

Application

Alkyne amino acid, more stable for peptide stapling via click chemistry than propargylglycine.

related product

Product No.
Description
Pricing

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service