Skip to Content
Merck
All Photos(1)

Documents

DWKW242832

Wheaton® Dropping Bottle

white bottle, capacity 6 mL

Synonym(s):

W242832

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

material

white bottle

packaging

pkg of 100 ea

manufacturer/tradename

WHEATON® W242832

capacity

6 mL

Looking for similar products? Visit Product Comparison Guide

Application

Opthalmic, IVD Kits, Other

Features and Benefits

  • Bottle made of natural or white LDPE (Low Density Polyethylene
  • White LDPE bottle helps protect UV light sensitive samples
  • Dropper tip caps and tips sold separately
  • Packaged in cases of 100 or 1000

Legal Information

WHEATON is a registered trademark of Wheaton Industries, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service