Skip to Content
Merck
All Photos(1)

Documents

CLS985025XX

Pyrex® flat bottom culture tube, rimless

capacity 59.0 mL

Synonym(s):

culture tubes, test tubes, Corning® 9850

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121706
NACRES:
NB.43

material

borosilicate glass
flat bottom tube

sterility

non-sterile

feature

cap: no

packaging

pack of 36 ea
case of 72

manufacturer/tradename

Corning 9850-25XX

capacity

59.0 mL

diam. × L

25 mm × 150 mm

color

clear

suitability

suitable for (plant culture)

Looking for similar products? Visit Product Comparison Guide

General description

Ideal for plant tissue culture and general culture work. The flat bottom facilitates use with agar slants.

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service