Skip to Content
Merck
All Photos(1)

Documents

CLS615001L

PYREXPLUS® narrow-mouth reagent bottle, with Pyrex® ST stopper, protective coating

capacity 1000 mL

Synonym(s):

Corning® 61500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

borosilicate glass bottle
clear bottle
cylindrical bottle
narrow-mouth bottle

stopper size

29

feature

autoclavable
cap

manufacturer/tradename

Corning 61500-1L

capacity

1000 mL

diam. × H

107 mm × 200.5 mm

Looking for similar products? Visit Product Comparison Guide

General description

Features a protective plastic coating that helps prevent glass from shattering and reduces any spills. Fully autoclavable (121°C) and resistant to thermal shock. Bottle has a ground neck,

Legal Information

Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
PYREXPLUS is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service