Key Documents
CLS5000125
Pyrex® wide-mouth Erlenmeyer flask with joint
capacity 125 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer, Corning® 5000
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Pricing and availability is not currently available.
material
borosilicate glass
bottom (baffled)
glass cap
narrow-mouth
Joint
joint ST/NS24/40
sterility
non-sterile
feature
cap
closure type Standard Taper joint
packaging
case of 12
pack of 6
manufacturer/tradename
Corning 5000-125
capacity
125 mL
General description
  • Pyrex® Erlenmeyer flasks are made of borosilicate glass for optical clarity and mechanical strength.
  • Flasks have durable white enamel graduations to show approximate capacity.
  • Large marking spot for labeling.
Graduated to show approximate capacity. Ref. ASTM E123.
Other Notes
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)