Key Documents
CLS1373160
Corning® wide mouth graduated milk dilution bottle
capacity 160 mL, screw cap
Synonym(s):
storage vessel
About This Item
UNSPSC Code:
41121800
Pricing and availability is not currently available.
material
borosilicate glass bottle
cylindrical bottle
phenolic cap
feature
autoclavable
cap
graduations
packaging
pack of 12 ea
case of 48 ea
manufacturer/tradename
Corning 1373-160
bottle neck diam.
~72 mm
capacity
160 mL
General description
Wide mouth bottle for easy measuring or filling. With cut-line graduation mark at 99 ± 1mL. Phenolic cap with rubber liner will withstand sterilization (wet at 121°C).
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)