BR780702
BRAND® micro tube with attached screw cap and sealing cone
capacity 1.5 mL, conical bottom, non-sterile
Synonym(s):
Micro Tubes with Screw caps, Microcentrifuge Tubes, Screw caps, Tubes
All Photos(1)
About This Item
Recommended Products
material
colorless cap
colorless tube
conical bottom
polyethylene cap
polypropylene tube
screw top cap
sterility
non-sterile
feature
graduations: no
packaging
pack of 1000 ea
manufacturer/tradename
BRAND 780702
capacity
1.5 mL
tube diam.
13 mm
General description
BRAND reaction tubes with attached cover (PE) reduce the danger of contamination or mistakes due to mixing up covers. The sealing cone of the cover lies seamlessly against the interior of the tube, preventing the contents from coming into contact with the thread.Application: -90 °C to +100 °CAttached covers are suitable for colored cover inserts. BIO-CERT® PCR QUALITY: Free of human DNA, RNase, DNase, Pyrogens and PCR-Inhibitors
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG