Skip to Content
Merck
All Photos(1)

Documents

BR705201

BRAND® HandyStep touch with universal AC adaptor and charging stand

Synonym(s):

Charging Stand, Electronic Pipette, HandyStep, HandyStep Touch, Positive Displacement Pipette, Repeater, Repeater Pipette, Repetitive Pipette

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121506
NACRES:
NB.01

packaging

pack of 1 ea

manufacturer/tradename

BRAND 705201

Related Categories

General description

Repetitive pipettes are the first choice when it comes to efficient dispensing of long series. Positive displacement pipettes are often the right solution for pipetting viscous media. The new stepper HandyStep® touch from BRAND combines both features.

With the touchscreen display, you conveniently swipe back and forth between functions and always have an overview of the essential information on your work steps. Operation is easy, even with protective gloves on. And speaking of easy, the days of manually changing tips are over. On the HandyStep® touch, lightly tapping on the Eject icon on the display is enough to eject the tip.

The HandyStep® touch repetitive pipette automatically recognizes the volume of the new BRAND PD- Tips II. This saves time and reduces the risk of error. Tips from many other manufacturers can also be used, providing maximum flexibility.
Charging is easy and efficient: simply place the HandyStep® touch in the inductive charging stand and the battery automatically starts charging.

Overview
  • Touchscreen operation with intuitive menu navigation in English, German, Spanish, French and Chinese
  • HandyStep touch with the features multi-dispensing, auto-dispensing and pipetting; HandyStep touch S with the additional features sequential dispensing, multi-aspiration and titration
  • Automatic size detection of BRAND PD-Tips II
  • Compatible with PD-Tips prior to the PD-Tips II generation and with tips from many other manufacturers: for example, Encode tips, Repet-Tips, Combitips, Combitips plus, Combitips advanced (the above-mentioned brands are property of their respective owners)
  • Automatic tip ejection
  • Variable volume adjustment from 1.0 μl to 50 ml
  • Easy to clean thanks to smooth surface without grooves and edges
  • Inductively rechargeable

Application

auto-pipetting, multi-dispensing

Features and Benefits

Overview:
  • Touchscreen operation with intuitive menu navigation in English, German, Spanish, French and Chinese
  • Multi-dispensing, auto-dispensing and pipetting
  • Automatic size detection of BRAND PD-Tips II
  • Compatible with PD-Tips prior to the PD-Tips II generation and with tips from many other manufacturers: for example, Encode tips, Repet-Tips, Combitips®, Combitips plus®, Combitips advanced® (the above-mentioned brands are property of their respective owners)
  • Automatic tip ejection
  • Variable volume adjustment from 1.0 µl to 50 ml
  • Easy to clean thanks to smooth surface without grooves and edges
  • Inductively rechargeable


Limitations of use:
  • Operating temperature of instrument and reagent should be between +15 °C and +40 °C (59 °F to 104 °F)
  • Vapor pressure up to 500 mbar
  • Viscosity: At 50 ml PD-Tip: 20 mPa s; at 5 ml PD-Tip: 260 mPa s; at 1,25 ml PD-Tip: 977 mPa s

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
Combitips is a registered trademark of Eppendorf AG
Combitips advanced is a registered trademark of Eppendorf AG
Combitips plus is a registered trademark of Eppendorf AG
HandyStep is a registered trademark of BRAND GMBH + CO KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service