Key Documents
BAF420120000
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
volume 0.09 ft3
Synonym(s):
SP Bel-Art® "Space Saver" Polycarbonate Vacuum Desiccator with Clear Polycarbonate Bottom
About This Item
UNSPSC Code:
41104510
NACRES:
NB.43
Pricing and availability is not currently available.
material
clear polycarbonate
packaging
pkg of 1 ea
manufacturer/tradename
Bel-Art F42012-0000
volume
0.09 ft3
General description
Degas liquids quickly and easily in the laboratory or classroom with this small plastic vacuum desiccator. These strong, shatterproof units provide dry storage and protection for sensitive products.
  • Lighter and less costly than similarly sized glass models; does not chip like glass
  • Holds a full vacuum (29.9" Hg) at room temperature for 24 hours
  • Tapered polypropylene stopcock with PTFE plug accepts tubing with ¼ to ⅜” I.D. and allows a controlled entrance of air preventing unnecessary turbulence
  • Neoprene O-ring assures a tight seal down to 0 ˚C without the need for grease (ring will not be harmed if grease is used)
  • ‘Flat Dome,’ clear polycarbonate top with large ribbed knob maximizes interior clearances and provides an average of 13% greater interior volume over standard hemispherical domes
  • All models include a 0.32 cm (⅛") thick perforated plate; disposable or reusable desiccant can be placed on or under this plate
  • White polypropylene bottom models can support incandescent crucibles on Minerit HD High Heat Ceramic Desiccator Plates
  • Clear polycarbonate bottom models offer increased viewing visibility from more angles
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)