Documents
BAF118590010
SP Bel-Art® jerrican with spigot
capacity 5 L (1.25 gal), high-density polyethylene
Synonym(s):
jerrycan, portable dispensor
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
material
high-density polyethylene
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F11859-0010
capacity
5 L (1.25 gal)
General description
Space-saving jerricans have a smooth turning spigot that opens and closes with half a turn for easy convenient dispensing from a stationary position. Made of high-density polyethylene, they have a unique 20cm (8") long spout with a built-in air vent. The 5 gallon jerrican has a 2.5cm (1") I.D. pouring spout; other sizes have 1.9cm (¾") I.D. spouts.
Features and Benefits
  • Great for dispensing without moving
  • Spout retracts into the threaded collar to conserve space
  • Collar is sealed by leakproof screw cap
  • Extra hand grip on the bottom gives positive control when pouring
  • Extra headspace for expansion of contents
  • Graduated in U.S. gallons and liters
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP