Skip to Content
Merck
All Photos(2)

Documents

B7252

Sigma-Aldrich

1-Bromo-2-naphthoic acid

≥98%

Synonym(s):

1-Bromo-2-naphthalenecarboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H7BrO2
CAS Number:
Molecular Weight:
251.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98%

form

powder

SMILES string

OC(=O)c1ccc2ccccc2c1Br

InChI

1S/C11H7BrO2/c12-10-8-4-2-1-3-7(8)5-6-9(10)11(13)14/h1-6H,(H,13,14)

InChI key

VUVIRKAVBZITDO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in synthesis of:
  • Binaphthyl-based amino acids and amino alcohols via domino coupling reactions and lactam ring opening of intermediates
  • Benzimidazoles and benzimidazolequinone derivatives via cyclocondensation
  • Axially chiral biaryls via Suzuki-Miyaura reactions
  • Aryl ring-fused benzimidazolequinones via radical cyclization
  • Probes for human cytochrome P450 enzymes
  • Phananthridinone derivatives via domino coupling reactions

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service