Skip to Content
Merck
All Photos(1)

Key Documents

B1126

Sigma-Aldrich

5-Benzyloxyindole-2-carboxylic acid ethyl ester

Synonym(s):

5-Benzyloxy-2-carbethoxyindole, NSC 30931

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO3
CAS Number:
Molecular Weight:
295.33
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

form

powder

storage temp.

−20°C

SMILES string

CCOC(=O)c1cc2cc(OCc3ccccc3)ccc2[nH]1

InChI

1S/C18H17NO3/c1-2-21-18(20)17-11-14-10-15(8-9-16(14)19-17)22-12-13-6-4-3-5-7-13/h3-11,19H,2,12H2,1H3

InChI key

DCIFXYFKVKDOLL-UHFFFAOYSA-N

Application

  • Reactant for preparation of indolyl ethanones as indoleamine 2,3-dioxygenase inhibitors
  • Reactant for preparation of histamine H3 receptor inverse agonists
  • Reactant for preparation of PPAR-γ binding agents with potential application to the treatment of osteoporosis
  • Reactant for preparation of Nonpeptide glycoprotein IIB/IIIA inhibitors

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service