Skip to Content
Merck
All Photos(1)

Documents

ALD00412

Sigma-Aldrich

Ethyl 4-(fluorosulfonyloxy)benzoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9FO5S
Molecular Weight:
248.23
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

description

Fp: >230°F

Quality Level

form

liquid

refractive index

n20/D 1.479

density

1.340 g/mL at 25 °C

SMILES string

O=S(OC1=CC=C(C(OCC)=O)C=C1)(F)=O

InChI

1S/C9H9FO5S/c1-2-14-9(11)7-3-5-8(6-4-7)15-16(10,12)13/h3-6H,2H2,1H3

InChI key

WSJHOCQDJHFOCL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The following aryl fluorosulfate can be utilized as a cross-coupling partner in Suzuki-Miyaura reactions. The standard reaction conditions for these palladium-catalyzed C-C bond formations are ligand-free and can be performed in water, at room temperatre and are not air sensitive.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service