Skip to Content
Merck
All Photos(1)

Key Documents

932833

Sigma-Aldrich

Tweezer

High Precision, Style 5, Locking

Synonym(s):

Gripper, Tweezer

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41111761
NACRES:
NB.35
Pricing and availability is not currently available.

General description

EMS techware

Application

High Precision and Ultra Fine tweezer with locking mechanism. Typical applications include tweezers for the electronic industry, watch-makers, jewelers and laboratory applications in moderately aggressive chemical environments. Straight, extra fine tips, superior finish, with locking system. Length: 110mm. The tweezers are the Swiss highest quality. High precision locking tweezer. Antimagnetic/anti-acid stainless steel. Superior finish.
Material: SA Steel . OAL: 110 mm. Tips: Extra fine with superior finish.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service