Skip to Content
Merck
All Photos(1)

Key Documents

929913

Sigma-Aldrich

24R,25-Dihydroxyvitamin D3-26,26,26,27,27,27-d6

≥97 atom % D, ≥98% (CP)

Synonym(s):

(3R,6R)-2-(Methyl-d3)-6-[(1R,3aS,4E,7aR)-octahydro-4-[(2Z)-2-[(5S)-5-hydroxy-2-methylenecyclohexylidene]ethylidene]-7a-methyl-1H-inden-1-yl]-2,3-heptane-1,1,1-d3-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27D6H38O3
Molecular Weight:
422.67
UNSPSC Code:
12142200
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥97 atom % D

Quality Level

Assay

≥98% (CP)

mass shift

M+6

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service