Skip to Content
Merck
All Photos(1)

Key Documents

92458

Sigma-Aldrich

Trimethylene sulfide

≥96.0% (GC)

Synonym(s):

Thietane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6S
CAS Number:
Molecular Weight:
74.14
Beilstein:
102383
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96.0% (GC)

form

liquid

refractive index

n20/D 1.510 (lit.)
n20/D 1.510

bp

94-94.5 °C (lit.)

density

1.028 g/mL at 25 °C (lit.)

SMILES string

C1CSC1

InChI

1S/C3H6S/c1-2-4-3-1/h1-3H2

InChI key

XSROQCDVUIHRSI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-4.0 °F - closed cup

Flash Point(C)

-20 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yu Ran Lee et al.
The journal of physical chemistry. A, 121(5), 1163-1167 (2017-01-15)
We have measured the high-resolution vibrational spectra of a thietane (trimethylene sulfide) cation in the gas phase by employing the vacuum ultraviolet mass-analyzed threshold ionization (VUV-MATI) spectroscopic technique. Peaks in the low-frequency region of the observed MATI spectrum of thietane
Rabi A Musah et al.
Plant physiology, 170(2), 1075-1089 (2015-12-15)
The roots of the shy plant Mimosa pudica emit a cocktail of small organic and inorganic sulfur compounds and reactive intermediates into the environment, including SO2, methanesulfinic acid, pyruvic acid, lactic acid, ethanesulfinic acid, propanesulfenic acid, 2-aminothiophenol, S-propyl propane 1-thiosulfinate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service