Skip to Content
Merck
All Photos(1)

Documents

906425

Sigma-Aldrich

DLAM-LVproR-13C4 Methyl Labeling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.12

technique(s)

bio NMR: suitable

Quality Level

shipped in

dry ice

storage temp.

−70°C

Related Categories

General description

DLAM-LVproR-13C4 , Kit has 13C4 isotopomer precursors and contains protocol instructions for creation of isotopically-labeled proteins.

Application

DLAM-LVproR-13C4 methyl labeling kit is used for stereospecific labeling of leucine and valine. This kit has been tested with protein isotopic labeling in E. coli. It is used to produce methyl-labeled proteins and acquire precise dynamic information with high resolution in solid-state NMR spectroscopy.
For protein methyl group assignment by 13C isotope labeling of amino acid methyl groups separately or simultaneously.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dae-Won Sim et al.
Molecules (Basel, Switzerland), 22(8) (2017-08-16)
A large portion of proteins in living organisms are membrane proteins which play critical roles in the biology of the cell, from maintenance of the biological membrane integrity to communication of cells with their surroundings. To understand their mechanism of
Rime Kerfah et al.
Journal of biomolecular NMR, 63(4), 389-402 (2015-11-15)
A new strategy for the NMR assignment of aliphatic side-chains in large perdeuterated proteins is proposed. It involves an alternative isotopic labeling protocol, the use of an out-and-back (13)C-(13)C TOCSY experiment ((H)C-TOCSY-C-TOCSY-(C)H) and an optimized non-uniform sampling protocol. It has
Silke Wiesner et al.
Current opinion in structural biology, 35, 60-67 (2015-09-26)
Intermolecular interactions are indispensible for biological function. Here we discuss how novel NMR techniques can provide unique insights into the assembly, dynamics and regulation of biomolecular complexes. We focus on applications that exploit the methyl TROSY effect and show that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service