Skip to Content
Merck
All Photos(2)

Key Documents

901216

Sigma-Aldrich

(S)-Ph-quinox

≥95%

Synonym(s):

(S)-4-Phenyl-2-(quinolin-2-yl)-4,5-dihydrooxazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H14N2O
CAS Number:
Molecular Weight:
274.32
UNSPSC Code:
12352200

Assay

≥95%

form

powder or crystals

reaction suitability

reagent type: catalyst
reagent type: ligand
reaction type: C-H Activation

mp

142 °C

Application

(S)-Ph-quinox is a chiral dinitrogen ligand which is used in the iridium-catalyzed enantioselective borylation[1] and silylation[2] of aromatic C-H bonds, in addition to the enantioselective palladium-catalyzed diamination of alkenes.[3] It can also be used in the aza-Wacker-type cyclization reaction of the olefinic tosylamides with molecular oxygen.[4]

related product

Product No.
Description
Pricing

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective Borylation of Aromatic C? H Bonds with Chiral Dinitrogen Ligands.
Su B, et al.
Angewandte Chemie (International Edition in English), 56(25), 7205-7208 (2017)
Pd-catalyzed asymmetric aza-Wacker-type cyclization reaction of olefinic tosylamides.
Jiang F, et al.
Tetrahedron Letters, 51(39), 5124-5126 (2010)
A Chiral Nitrogen Ligand for Enantioselective, Iridium?Catalyzed Silylation of Aromatic C? H Bonds.
Su B, et al.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 129(4), 1112-1116 (2017)
Erica L Ingalls et al.
Journal of the American Chemical Society, 135(24), 8854-8856 (2013-06-06)
An enantioselective Pd-catalyzed vicinal diamination of unactivated alkenes using N-fluorobenzenesulfonimide as both an oxidant and a source of nitrogen is reported. The use of Ph-pybox and Ph-quinox ligands afforded differentially protected vicinal diamines in good yields with high enantioselectivities. Mechanistic

Articles

QuinoxP*: Air-Stable and Highly Efficient and Productive Chiral Ligands

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service