Skip to Content
Merck
All Photos(2)

Key Documents

900975

Sigma-Aldrich

2-(4-Chloro-3-iodo-6-methyl-1H-pyrrolo[3,2-c]pyridin-1-yl)ethan-1-amine dihydrochloride

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11ClIN3 · 2HCl
Molecular Weight:
408.49
UNSPSC Code:
12352101
NACRES:
NA.22

Assay

≥95%

form

powder or crystals

functional group

amine
chloro
iodo

storage temp.

2-8°C

Application

Pyrrolo[3,2-c] pyridine derivative (5-azaindole) that can be used as an indole bioisostere for designing azaindole-based drugs.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of tetrahydro-5-azaindoles and 5-azaindoles using Pictet?Spengler reaction?appreciable difference in products using different acid catalysts.
Shumaila A MA, et al.
Tetrahedron, 67(5), 936-942 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service