900153
UV curable resin
adhesible to plastics and glass, for protective coatings, optical, and outdoor applications
Synonym(s):
CPS 1020 UV-A
About This Item
Recommended Products
form
liquid
viscosity
310 cP (Brookfield LDTDV-II digital viscometer with size 31 spindle)
density
1.16 g/mL
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
- The storage temperature should not exceed 35°C.
- Keep tightly closed. Store in a cool, dry, well-ventilated area.
- Do not expose to ultraviolet light.
- High temperatures (>80°C), pressure and contamination with peroxides may result in auto-polymerization of the product.
- Unstable upon depletion of inhibitor.
Application
- Cures rapidly to form a glassy, high modulus, optically clear material.
- Excellent for use in protective coatings, optical, and outdoor applications.
- Non-yellowing.
- Low oxygen inhibition.
- Mechanical properties after curing:
Strength*: 29.3 MPa
Elongation*: 3.4%
Tack Free**: 50 fpm
*Test Resources: tensile tester, 35 mm gauge length, 1.0 mm thickness, 6.5 mm width. ** Cured under stated intensity. Medium pressure mercury bulb with 320-500 nm filter. Adhesion tests were conducted using a cross hatch tape test on films 0.125 mm thick on glass and PET. Material data using ASTM D 638-02a.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service