Skip to Content
Merck
All Photos(1)

Key Documents

87778

Sigma-Aldrich

Dansyl-L-proline

≥99.0% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O4S
CAS Number:
Molecular Weight:
348.42
Beilstein:
497317
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥99.0% (TLC)

optical activity

[α]20/D −45±2°, c = 1% in 1 M HCl

mp

150-153 °C

storage temp.

2-8°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)N3CCC[C@H]3C(O)=O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Derivatizing agent for HPLC; assay of enantiomeric purity of amines[1]

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K. Gamoh, H. Sawamoto
Analytica Chimica Acta, 243, 251-251 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service