Skip to Content
Merck
All Photos(2)

Key Documents

80940

Sigma-Aldrich

Ethyl pivaloylacetate

≥97.0% (GC)

Synonym(s):

4,4-Dimethyl-3-oxopentanoic acid ethyl ester, Ethyl 4,4-dimethyl-3-oxopentanoate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CCOCH2COOC2H5
CAS Number:
Molecular Weight:
172.22
Beilstein:
1767555
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

refractive index

n20/D 1.432

bp

88-90 °C/10 mmHg (lit.)

density

0.967 g/mL at 20 °C (lit.)

functional group

ester
ketone

SMILES string

CCOC(=O)CC(=O)C(C)(C)C

InChI

1S/C9H16O3/c1-5-12-8(11)6-7(10)9(2,3)4/h5-6H2,1-4H3

InChI key

VUYNTIDSHCJIKF-UHFFFAOYSA-N

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

172.4 °F - closed cup

Flash Point(C)

78 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service