Skip to Content
Merck
All Photos(1)

Key Documents

797243

Sigma-Aldrich

4,7-Dibromo-5,6-difluoro-2,1,3-benzothiadiazole

Synonym(s):

bb-F2BT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6Br2F2N2S
CAS Number:
Molecular Weight:
329.95
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97% (HPLC)

Quality Level

form

fibers

mp

150-155 °C

storage temp.

2-8°C

SMILES string

FC1=C(Br)C2=NSN=C2C(Br)=C1F

InChI

1S/C6Br2F2N2S/c7-1-3(9)4(10)2(8)6-5(1)11-13-12-6

InChI key

HFUBKQHDPJZQIW-UHFFFAOYSA-N

Application

Fluorinated benzothiadiazole-based conjugated polymers for high open-circuit voltage in organic photovoltaics.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silaindacenodithiophene-Based Low Band Gap Polymers - The Effect of Fluorine Substitution on Device Performances and Film Morphologies
Schroeder, Bob C.; Huang, Zhenggang; Ashraf, Raja Shahid; Smith, Jeremy; et al
Advances in Functional Materials, 22(8), 1663-1670 (2012)
Jonggi Kim et al.
ACS applied materials & interfaces, 6(10), 7523-7534 (2014-04-22)
The introduction of fluorine (F) atoms onto conjugated polymer backbone has verified to be an effective way to enhance the overall performance of polymer-based bulk-heterojunction (BHJ) solar cells, but the underlying working principles are not yet fully uncovered. As our
Yong Zhang et al.
Chemical communications (Cambridge, England), 47(39), 11026-11028 (2011-09-13)
Small band-gap conjugated polymers based on monofluoro- and difluoro-substituted benzothiadiazole were developed. Highly efficient polymer solar cells (PCE as high as 5.40%) could be achieved for devices made from these polymers.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service