Skip to Content
Merck
All Photos(1)

Key Documents

779598

Sigma-Aldrich

Methyl 4-azidobenzoate solution

~0.5 M in tert-butyl methyl ether, ≥95% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H7N3O2
CAS Number:
Molecular Weight:
177.16
Beilstein:
1960489
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

solution

concentration

~0.5 M in tert-butyl methyl ether

impurities

≤2.0% water

storage temp.

−20°C

SMILES string

COC(=O)c1ccc(cc1)N=[N+]=[N-]

InChI

1S/C8H7N3O2/c1-13-8(12)6-2-4-7(5-3-6)10-11-9/h2-5H,1H3

InChI key

RULRGDSQDQHZOK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT RE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-27.4 °F

Flash Point(C)

-33 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemoselective reduction of azides catalyzed by molybdenum xanthate by using phenylsilane as the hydride source.
Maddani, M. R., et al.
Tetrahedron, 66, 329-333 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service