Skip to Content
Merck
All Photos(1)

Documents

774952

Sigma-Aldrich

Furfuryl alcohol-alpha-13C

≥99 atom % 13C, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
13CC4H6O2
Molecular Weight:
99.09
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥97% (CP)

form

liquid

mass shift

M+1

SMILES string

O[13CH2]C1=CC=CO1

InChI

1S/C5H6O2/c6-4-5-2-1-3-7-5/h1-3,6H,4H2/i4+1

InChI key

XPFVYQJUAUNWIW-AZXPZELESA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 2 - Eye Irrit. 2 - STOT RE 2 Inhalation - STOT SE 3

Target Organs

Nose, Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

149.0 °F - closed cup

Flash Point(C)

65 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service