760455
Ethyl 2-(phenylcarbonothioylthio)propionate
97% (HPLC)
Synonym(s):
2-[(Phenylthioxomethyl)thio]propanoic acid ethyl ester, Ethyl 2-(phenylcarbonothioylthio)propanoate
About This Item
Assay
97% (HPLC)
form
liquid
refractive index
n20/D 1.611
density
1.163 g/mL at 25 °C
storage temp.
2-8°C
SMILES string
CCOC(=O)C(C)SC(=S)c1ccccc1
InChI
1S/C12H14O2S2/c1-3-14-11(13)9(2)16-12(15)10-7-5-4-6-8-10/h4-9H,3H2,1-2H3
InChI key
NXWVPIWRMPQYEX-UHFFFAOYSA-N
General description
Application
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
The modification of biomacromolecules, such as peptides and proteins, through the attachment of synthetic polymers has led to a new family of highly advanced biomaterials with enhanced properties.
Micro review of reversible addition/fragmentation chain transfer (RAFT) polymerization.
Protocols
We present an article about RAFT, or Reversible Addition/Fragmentation Chain Transfer, which is a form of living radical polymerization.
We presents an article featuring procedures that describe polymerization of methyl methacrylate and vinyl acetate homopolymers and a block copolymer as performed by researchers at CSIRO.
Polymerization via ATRP procedures demonstrated by Prof. Dave Haddleton's research group at the University of Warwick.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service