Skip to Content
Merck
All Photos(1)

Key Documents

760447

Sigma-Aldrich

10-Undecenyl acrylate

contains <1000 ppm MEHQ as inhibitor, 98%

Synonym(s):

2-Propenoic acid, undec-10-enyl ester, Undec-10-enyl prop-2-enoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H24O2
CAS Number:
Molecular Weight:
224.34
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

liquid

contains

<1000 ppm MEHQ as inhibitor

refractive index

n20/D 1.451

density

0.882 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

C=CCCCCCCCCOC(=O)C=C

InChI

1S/C13H22O2/c1-3-5-6-7-8-9-10-11-12-15-13(14)4-2/h3-4H,1-2,5-12H2

InChI key

WDGPKCJWRJSYLS-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service