Skip to Content
Merck
All Photos(1)

Key Documents

75095

Sigma-Aldrich

Oleic anhydride

≥95.0% (GC)

Synonym(s):

cis-9-Octadecenoic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)7CH=CH(CH2)7CO]2O
CAS Number:
Molecular Weight:
546.91
Beilstein:
1730211
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (GC)

form

liquid

bp

200-215 °C/11 mmHg (lit.)

mp

22-24 °C (lit.)

SMILES string

CCCCCCCC\C=C/CCCCCCCC(=O)OC(=O)CCCCCCC\C=C/CCCCCCCC

InChI

1S/C36H66O3/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-35(37)39-36(38)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h17-20H,3-16,21-34H2,1-2H3/b19-17-,20-18-

InChI key

OCNZHGHKKQOQCZ-CLFAGFIQSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna Hrabovska et al.
PloS one, 5(9), e12892-e12892 (2010-10-05)
We wished to develop alternate production strategies to generate antibodies against traditionally problematic antigens. As a model we chose butyrylcholinesterase (BChE), involved in termination of cholinergic signaling, and widely considered as a poor immunogen. Jettisoning traditional laborious in silico searching
Bhaswati Sengupta et al.
Journal of lipid research, 54(12), 3358-3372 (2013-10-12)
Generation of foam cells, an essential step for reverse cholesterol transport studies, uses the technique of receptor-dependent macrophage loading with radiolabeled acetylated LDL. In this study, we used the ability of a biologically relevant detergent molecule, lysophosphatidylcholine (lyso-PtdCho), to form
Sara García-Linares et al.
Biochemistry, 55(48), 6630-6641 (2016-12-10)
Sea anemone actinoporins constitute a protein family of multigene pore-forming toxins (PFT). Equinatoxin II (EqtII), fragaceatoxin C (FraC), and sticholysins I and II (StnI and StnII, respectively), produced by three different sea anemone species, are the only actinoporins whose molecular
Eliete J B Bighetti et al.
Lipids in health and disease, 8, 50-50 (2009-11-26)
CETP is a plasma protein that modulates atherosclerosis risk through its HDL-cholesterol reducing action. The aim of this work was to examine the effect of the PPARalpha agonist, ciprofibrate, on the CETP gene expression, in the presence and absence of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service