Skip to Content
Merck
All Photos(1)

Key Documents

742228

Sigma-Aldrich

Ethyl (E)-3-(1-pyrrolidinyl)crotonate

99% (GC)

Synonym(s):

Ethyl (E)-3-(1-pyrrolidinyl)-2-butenoate, Ethyl (E)-3-pyrrolidinocrotonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO2
CAS Number:
Molecular Weight:
183.25
Beilstein:
1074392
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99% (GC)

form

liquid

suitability

complies for identity (IR)

functional group

ester

SMILES string

CCOC(=O)\C=C(/C)N1CCCC1

InChI

1S/C10H17NO2/c1-3-13-10(12)8-9(2)11-6-4-5-7-11/h8H,3-7H2,1-2H3/b9-8+

InChI key

MSOQKPXSIHLODG-CMDGGOBGSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service