Skip to Content
Merck
All Photos(2)

Key Documents

738972

Sigma-Aldrich

5-Amino-3-bromo-2-chloropyridine

97%

Synonym(s):

3-Amino-5-bromo-6-chloropyridine, 5-Bromo-6-chloro-3-pyridinamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4BrClN2
CAS Number:
Molecular Weight:
207.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

118-123 °C

SMILES string

Nc1cnc(Cl)c(Br)c1

InChI

1S/C5H4BrClN2/c6-4-1-3(8)2-9-5(4)7/h1-2H,8H2

InChI key

ISKBXMMELYRESC-UHFFFAOYSA-N

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service