Skip to Content
Merck
All Photos(2)

Documents

728926

Sigma-Aldrich

2-Amino-5-phenylpyrazine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9N3
CAS Number:
Molecular Weight:
171.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

powder

mp

143-148 °C

SMILES string

Nc1cnc(cn1)-c2ccccc2

InChI

1S/C10H9N3/c11-10-7-12-9(6-13-10)8-4-2-1-3-5-8/h1-7H,(H2,11,13)

InChI key

KJAKXVBZQBPPOB-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service