Skip to Content
Merck
All Photos(1)

Documents

725404

Sigma-Aldrich

Bis(2-(bis(diethylamino)phosphino)phenyl) ether

95%

Synonym(s):

Bis(2-(bis(diethylamido)phosphino)phenyl) ether

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C28H48N4OP2
CAS Number:
Molecular Weight:
518.65
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Assay

95%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.565

functional group

phosphine

storage temp.

2-8°C

SMILES string

CCN(CC)P(N(CC)CC)c1ccccc1Oc2ccccc2P(N(CC)CC)N(CC)CC

InChI

1S/C28H48N4OP2/c1-9-29(10-2)34(30(11-3)12-4)27-23-19-17-21-25(27)33-26-22-18-20-24-28(26)35(31(13-5)14-6)32(15-7)16-8/h17-24H,9-16H2,1-8H3

InChI key

XYBRYIYICOWBBB-UHFFFAOYSA-N

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service