Skip to Content
Merck
All Photos(2)

Key Documents

725080

Sigma-Aldrich

5-Ethyl-2-methylpyridine borane complex

Synonym(s):

(5-Ethyl-2-methylpyridine)trihydroborane, 5-Ethyl-2-methylpyridine borane, PEMB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14BN
CAS Number:
Molecular Weight:
135.01
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reagent type: reductant

density

0.907 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

B.CCc1ccc(C)nc1

InChI

1S/C8H11N.BH3/c1-3-8-5-4-7(2)9-6-8;/h4-6H,3H2,1-2H3;1H3

InChI key

MNMBLEICNIBHKB-UHFFFAOYSA-N

Application

Reactant for:
  • Organometallic cross-coupling reactions
  • Reductive amination

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

221.0 °F

Flash Point(C)

105 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service