Skip to Content
Merck
All Photos(1)

Key Documents

72251

Sigma-Aldrich

Nickel(II) carbonate basic hydrate

purum p.a., 48-50% Ni basis (KT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
~NiCO3 · 2Ni(OH)2 · 4H2O
CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

purum p.a.

form

powder

concentration

48-50% Ni (KT)

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤500 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤50 mg/kg
Na: ≤5000 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Ni++].O[Ni]O.O[Ni]O.[O-]C([O-])=O

InChI

1S/CH2O3.3Ni.4H2O/c2-1(3)4;;;;;;;/h(H2,2,3,4);;;;4*1H2/q;3*+2;;;;/p-6

InChI key

ATYNVYNPWGEMMI-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1A Inhalation - Muta. 2 - Repr. 1B - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service