Skip to Content
Merck
All Photos(3)

Key Documents

718823

Sigma-Aldrich

1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23BN2O3
CAS Number:
Molecular Weight:
278.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

74-78 °C

functional group

ether

storage temp.

2-8°C

SMILES string

CC1(C)OB(OC1(C)C)c2ccnn2C3CCCCO3

InChI

1S/C14H23BN2O3/c1-13(2)14(3,4)20-15(19-13)11-8-9-16-17(11)12-7-5-6-10-18-12/h8-9,12H,5-7,10H2,1-4H3

InChI key

ZZRFDLHBMBHJTI-UHFFFAOYSA-N

Application

1-(Tetrahydro-2H-pyran-2-yl)-1H-pyrazole-5-boronic acid pinacol ester can be used as a reactant:
  • In Suzuki coupling reaction for the preparation of heteroaryl scaffolds via formation of C-C bond.[1]
  • To synthesize darolutamide derivatives as potential androgen receptor inhibitors.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery and biological evaluation of darolutamide derivatives as inhibitors and down-regulators of wild-type AR and the mutants
Y Jiang, et al.
European Journal of Medicinal Chemistry, 182, 111608-111608 (2019)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service