Skip to Content
Merck
All Photos(2)

Key Documents

716707

Sigma-Aldrich

Cadmium zinc telluride

99.9999% trace metals basis

Synonym(s):

CZT

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cd0.9Zn0.1Te
Molecular Weight:
235.31
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9999% trace metals basis

form

crystals

mp

1100-1150 °C

SMILES string

[Zn]=[Te].[Cd]=[Te]

InChI

1S/Cd.2Te.Zn

InChI key

KHJSUZZLLOGDFZ-UHFFFAOYSA-N

General description

Cadmium zinc telluride (Cd1-xZnxTe or CZT) can detect nuclear radiation hence, CZT with Zn concentrations between 0.1-0.2 may be used in the making of radiation detectors of X-ray and gamma-ray spectrometers.[1][2][3][4]

Application

A direct bandgap semiconductor, it is used in a variety of applications, including radiation detectors, photorefractive gratings, electro-optic modulators, solar cells, and terahertz generation and detection. The band gap varies from approximately 1.4 to 2.2 eV, depending on composition.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabrina Koehler et al.
Frontiers in zoology, 10(1), 3-3 (2013-02-02)
Insects have evolved a wide range of mechanisms to defend themselves and their offspring against antagonists. One of these strategies involves the utilization of antimicrobial compounds provided by symbiotic bacteria to protect the host or its nutritional resources from pathogens
John A Kennedy et al.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology, 26(1), 236-245 (2017-05-04)
Data regarding cardiac cadmium-zinc-telluride (CZT)-specific augmented databases and their impact on CT-based attenuation correction (AC) perfusion scores in myocardial perfusion imaging (MPI) were obtained on a multiple-pinhole CZT SPECT/CT. Summed stress (SSS) and rest scores (SRS) were measured using automated
Catherine Gebhard et al.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology, 27(1), 159-170 (2018-04-25)
In light of growing cardiovascular mortality rates observed in young women, sexual dimorphism in cardiac autonomic nervous control is gaining increasing attention. Heart rate responses to adenosine mirror autonomic activity and may carry important prognostic information. Hemodynamic changes during adenosine
Alessia Gimelli et al.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology, 27(1), 71-79 (2017-01-14)
Single-photon emission computed-tomography (SPECT) allows the quantification of LV eccentricity index (EI), a measure of cardiac remodeling. We sought to evaluate the feasibility of EI measurement with SPECT myocardial perfusion imaging and its interactions with relevant LV functional and structural
Alessia Gimelli et al.
Journal of nuclear cardiology : official publication of the American Society of Nuclear Cardiology, 24(2), 687-695 (2016-02-06)
Myocardial perfusion imaging (MPI) performed on traditional single-photon emission computed-tomography cameras has been shown to have a sub-optimal accuracy in detecting multivessel coronary artery disease (CAD). Six-hundred and ninety-five patients were submitted to MPI on a novel cadmium-zinc-telluride (CZT) camera

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service