Skip to Content
Merck
All Photos(1)

Key Documents

715646

Sigma-Aldrich

N-Boc-3-(methylamino)piperidine

96%

Synonym(s):

3-(Methylamino)piperidine-1-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H22N2O2
CAS Number:
Molecular Weight:
214.30
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.467

density

0.983 g/cm3 at 25 °C

storage temp.

2-8°C

SMILES string

CNC1CCCN(C1)C(=O)OC(C)(C)C

InChI

1S/C11H22N2O2/c1-11(2,3)15-10(14)13-7-5-6-9(8-13)12-4/h9,12H,5-8H2,1-4H3

InChI key

XRRRUOWSHGFPTI-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • TRPV1 antagonists
  • Aminopiperidines as iNOS inhibitors

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service