Skip to Content
Merck
All Photos(1)

Key Documents

712353

Sigma-Aldrich

(2R,2′R)-1,1′-Bis(2-pyridinylmethyl)-2,2′-bipyrrolidine tetrahydrochloride

≥96.0%

Synonym(s):

(R,R′)-PDP tetrahydrochloride, (R,R)-BPBP tetrahydrochloride, 2,2′-[(2R,2′R)-[2,2′-Bipyrrolidine]-1,1′-diylbis(methylene)]bis-pyridine tetrahydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H26N4 · 4HCl
CAS Number:
Molecular Weight:
468.29
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Assay

≥96.0% (HPLC)
≥96.0%
96.0-104.0% (AT)

form

powder

optical purity

enantiomeric excess: ≥98.0%

SMILES string

Cl.Cl.Cl.Cl.C1C[C@H]([C@H]2CCCN2Cc3ccccn3)N(C1)Cc4ccccn4

InChI

1S/C20H26N4.4ClH/c1-3-11-21-17(7-1)15-23-13-5-9-19(23)20-10-6-14-24(20)16-18-8-2-4-12-22-18;;;;/h1-4,7-8,11-12,19-20H,5-6,9-10,13-16H2;4*1H/t19-,20-;;;;/m1..../s1

InChI key

WTQUEUGYNCZETF-XQHCZWOQSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuclear physics. A lab to get the measure of matter.
Daniel Clery
Science (New York, N.Y.), 318(5851), 738-739 (2007-11-03)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service