Skip to Content
Merck
All Photos(1)

Documents

712213

Sigma-Aldrich

Boc-Dab-OH

≥97.0%

Synonym(s):

(S)-4-Amino-2-(tert-butoxycarbonylamino)butanoic acid, Nα-Boc-L-2,4-diaminobutyric acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H18N2O4
CAS Number:
Molecular Weight:
218.25
Beilstein:
4745487
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥97.0% (TLC)
≥97.0%

optical activity

[α]/D +7.5±1°, c = 1 in methanol: water (1:1)

impurities

≤2.0% water

application(s)

peptide synthesis

SMILES string

CC(C)(C)OC(=O)N[C@@H](CCN)C(O)=O

InChI

1S/C9H18N2O4/c1-9(2,3)15-8(14)11-6(4-5-10)7(12)13/h6H,4-5,10H2,1-3H3,(H,11,14)(H,12,13)/t6-/m0/s1

InChI key

MDCPCLPRWLKUIQ-LURJTMIESA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service