Skip to Content
Merck
All Photos(3)

Key Documents

710024

Sigma-Aldrich

1-Phenylvinylboronic acid MIDA ester

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H14BNO4
CAS Number:
Molecular Weight:
259.07
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

186-190 °C

functional group

phenyl

SMILES string

CN1CC(=O)OB(OC(=O)C1)C(=C)c2ccccc2

InChI

1S/C13H14BNO4/c1-10(11-6-4-3-5-7-11)14-18-12(16)8-15(2)9-13(17)19-14/h3-7H,1,8-9H2,2H3

InChI key

LDAWJVVAQPYNSX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabin Llona-Minguez et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(20), 7394-7398 (2015-03-27)
A two-step synthesis of structurally diverse pyrrole-containing bicyclic systems is reported. ortho-Nitro-haloarenes coupled with vinylic N-methyliminodiacetic acid (MIDA) boronates generate ortho-vinyl-nitroarenes, which undergo a "metal-free" nitrene insertion, resulting in a new pyrrole ring. This novel synthetic approach has a wide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service