Skip to Content
Merck
All Photos(1)

Key Documents

703095

Sigma-Aldrich

Hydrogen[4-[bis(2,4,6-trimethylphenyl)phosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate

95%

Synonym(s):

Frustrated phosphonium borate 2, Hydrogen[4-[dimesitylphosphino]-2,3,5,6-tetrafluorophenyl]hydrobis(2,3,4,5,6-pentafluorophenyl)borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H+[(((CH3)3C6H2)2PC6F4)B(H) (C6F5)2]
CAS Number:
Molecular Weight:
764.34
MDL number:
UNSPSC Code:
12352300

Assay

95%

mp

211-216 °C

storage temp.

2-8°C

SMILES string

Cc1cc(C)c([PH+](c2c(C)cc(C)cc2C)c3c(F)c(F)c([BH-](c4c(F)c(F)c(F)c(F)c4F)c5c(F)c(F)c(F)c(F)c5F)c(F)c3F)c(C)c1

InChI

1S/C36H23BF14P/c1-11-7-13(3)34(14(4)8-11)52(35-15(5)9-12(2)10-16(35)6)36-32(50)24(42)19(25(43)33(36)51)37(17-20(38)26(44)30(48)27(45)21(17)39)18-22(40)28(46)31(49)29(47)23(18)41/h7-10,37H,1-6H3/q-1/p+1

InChI key

IENZHWXRYKZMGT-UHFFFAOYSA-O

Application

Catalyst for metal-free hydrogenation

Reactant for:
  • Formation of phosphine-boranes and cationic phosphonium-boranes
  • Metal-free hydrogen activation
Frustrated phosphonium borate for metal-free catalytic hydrogenation of imines. Activates dihydrogen under mild conditions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal-free catalytic hydrogenation.
Preston A Chase et al.
Angewandte Chemie (International ed. in English), 46(42), 8050-8053 (2007-08-19)
Gregory C Welch et al.
Science (New York, N.Y.), 314(5802), 1124-1126 (2006-11-18)
Although reversible covalent activation of molecular hydrogen (H2) is a common reaction at transition metal centers, it has proven elusive in compounds of the lighter elements. We report that the compound (C6H2Me3)2PH(C6F4)BH(C6F5)2 (Me, methyl), which we derived through an unusual

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service